Global Water Hydrant Water Pressure Logger
Shaped like a fire hydrant cap to reduce theft or vandalization
  • High-speed sampling mode captures momentary pressure events
  • USB data port and bundled software allow for easy setup, calibration, and data transfer
Easy-to-use, rugged pressure logger lets you quickly locate distribution pressure problems and gather system modeling data. Massive, nonvolatile memory stores over 81,000 pressure readings. High-speed sampling mode catches events like pressure spikes and water hammer. Flexible datalogging capabilities allow defined interval, high speed, and exception sampling. Bundled software lets user easily export readings via USB data port to a computer. Software additionally provides for easy setup, calibration, and data management.
$840.00 - $869.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Global Water PL200-H-2 Hydrant Water Pressure Logger with BSPP to NPTF Adapter EW-32922-80 Mfr # FUB000K
COMPARE
Hydrant Water Pressure Logger with BSPP to NPTF Adapter
$869.00USD / Each
Global Water PL200-H-1 hydrant water pressure logger EW-32922-82 Mfr # FUA000K
COMPARE
hydrant water pressure logger
$840.00USD / Each