GLI/Hach Differential Measurement pH and ORP Electrodes

Unsurpassed accuracy with differential measurement technique
  • Three electrodes instead of the normal two in conventional pH sensors
  • Greater reliability resulting in less downtime and maintenance
  • Replaceable salt bridge/protector
  • Encapsulated electronics and preamp
  • Versatile body styles and mounting hardware including retrofit
  • 914 m (3000 ft) transmission distance
Differential Electrode measurement technique uses three electrodes instead of the two normally used in conventional pH sensors. Process and reference electrodes measure the pH differentially with respect to a third ground electrode. The end result is unsurpassed measurement accuracy, reduced reference junction potential, and elimination of sensor ground loops. These sensors provide greater reliability, resulting in less downtime and maintenance. The unique, replaceable salt bridge holds an extraordinary volume of buffer to extend the working life of the sensor by protecting the reference electrode from harsh process conditions. The salt bridge simply threads onto the end of the sensor if replacement is needed. Encapsulated design of the preamp protects the sensor’s built-in preamp from moisture and humidity, ensuring reliable sensor operation. The preamp in the pH sensor produces a strong signal, enabling the sensor to be located up to 914 m (3000 ft.) from the analyzer.
$1,091.00 - $1,532.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach 6028P0 Differential pH electrode, 10-ft cable, LCP body EW-27400-00 Mfr # 6028P0
COMPARE
Differential pH electrode, 10-ft cable, LCP body
$1,432.00USD / Each
Hach PD1P1 Differential pH electrode, 1" NPT (M), 15-ft cable, PEEK body EW-27400-02 Mfr # PD1P1
COMPARE
Differential pH electrode, 15-ft cable, PEEK body
$1,184.00USD / Each
Hach 2028RO Differential ORP electrode, 10-ft cable, LCP body EW-27400-10 Mfr # 2028R0
COMPARE
Differential ORP electrode, 10-ft cable, LCP body
$1,532.00USD / Each
Hach PD1R1 Differential pH electrode, 15-ft cable, PPS body EW-27400-12 Mfr # PD1R1
COMPARE
Differential pH electrode, 15-ft cable, PPS body
$1,091.00USD / Each
Hach RD1P5 Differential ORP electrode, 15-ft cable, PEEK body EW-27400-14 Mfr # RD1P5
COMPARE
Differential ORP electrode, 15-ft cable, PEEK body
$1,243.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat