GHS Health Hazard Pictogram Labels

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Health Hazard pictogram indicates the following potential hazard: carcinogen, mutagenicity, reproductive toxicity, respiratory sensitizer, target organ toxicity, aspiration toxicity.
$16.60 - $41.10USD / Roll of 250

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Quantity
Availability Pricing
Accuform LZH609PS2 GHS Label, Health Hazard Pictogram, 1" x 1", adhesive paper, 250/roll EW-50713-16 Mfr # LZH609PS2
COMPARE
1 Adhesive Paper 250/roll
$16.60USD / Roll of 250
Accuform LZH609PS5 GHS Label, Health Hazard Pictogram, 1" x 1", adhesive paper, 500/roll EW-50713-17 Mfr # LZH609PS5
COMPARE
1 Adhesive Paper 500/roll
$26.50USD / Roll of 500
Accuform LZH609EV2 GHS Label, Health Hazard Pictogram, 1" x 1", adhesive poly, 250/roll EW-50713-18 Mfr # LZH609EV2
COMPARE
1 Adhesive Poly 250/roll
$25.70USD / Roll of 250
Accuform LZH609EV5 GHS Label, Health Hazard Pictogram, 1" x 1", adhesive poly, 500/roll EW-50713-19 Mfr # LZH609EV5
COMPARE
1 Adhesive Poly 500/roll
$41.10USD / Roll of 500
Accuform LZH619EV2 GHS Label, Health Hazard Pictogram, 2" x 2", adhesive poly, 250/roll EW-50713-22 Mfr # LZH619EV2
COMPARE
2 Adhesive Poly 250/roll
$30.40USD / Roll of 250
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat