+GF+ Signet Conductivity/Resistivity Electrodes

Coupled with +GF+ Signet measuring circuitry, a three decade measurement range reading is achieved without the need for troublesome electrode platinization
  • Flow-through design eliminates bubble entrapment and sediment build-up
  • Removable/reversible sensor fitting design
  • Built-in strain relief
  • Controlled surface finish ensures accuracy and repeatability
  • In-line or submersible mounting
+GF+ Signet Conductivity/Resistivity Electrodes are designed to provide versatile installation and accurate sensing across a very broad dynamic range. Platinum RTD (PT-1000) located within the electrode allows optimal temperature sensing.
$515.50 - $1,350.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GF Signet 3-2819-1 2819 Conductivity Electrode; SS, K=0.01, 3/4"NPT(M) EW-19905-49 Mfr # 3-2819-1
COMPARE
2819 Conductivity Electrode; SS, K=0.01, 3/4\"NPT(M)
$515.50USD / Each
GF Signet 3-2820-1 2820 Conductivity Electrode; SS, K=0.1, 3/4"NPT(M) EW-19905-50 Mfr # 3-2820-1
COMPARE
2820 Conductivity Electrode; SS, K=0.1, 3/4\"NPT(M)
$517.50USD / Each
GF Signet 3-2821-1 2821 Conductivity Electrode; SS, K=1.0, 3/4"NPT(M) EW-19905-51 Mfr # 3-2821-1
COMPARE
2821 Conductivity Electrode; SS, K=1.0, 3/4\"NPT(M)
$541.00USD / Each
GF Signet 3-2822-1 2822 Conductivity Electrode; SS, K=10.0, 3/4"NPT(M) EW-19905-52 Mfr # 3-2822-1
COMPARE
2822 Conductivity Electrode; SS, K=10.0, 3/4\"NPT(M)
$933.00USD / Each
GF Signet 3-2820-S1 2820 Conductivity Electrode; SS, K=0.1, 1.5" Tri-Clamp EW-56560-39 Mfr # 3-2820-S1
COMPARE
2820 Conductivity Electrode; SS, K=0.1, 1.5\" Tri-Clamp
$1,350.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat