+GF+® Signet 2724-2726 DryLoc® pH and ORP Electrodes
Unique DryLoc design provides maximum installation flexibility
  • Chemically resistant Ryton® body for use in dirty or corrosive environments
  • Easy retrofitting! Mounts to existing off-the-shelf and +GF+ Signet installation fittings
Electrodes use a unique foul-proof DryLoc connector with gold plated contacts for extended lifetime. Choice of flat pH, bulb pH, and flat ORP sensing elements provides for a broad range of uses. HF resistant versions are for use in applications where traces of hydrofluoric acid will attack standard glass. LC versions are designed for use in low conductivity fluids and ultrapure water applications.
$385.25 - $735.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GF Signet 3-2724-00 pH Electrode, Flat Surface, Pt-1000 ATC, 3/4" NPT Threads
Always in Stock
EW-56561-00 Mfr # 3-2724-00
COMPARE
pH Electrode, flat surface, Pt-1000 ATC, 3/4\" NPT threads
$501.50USD / Each
GF Signet 3-2724-01 pH Electrode, Flat Surface, Pt-1000 ATC, ISO 7/1 R3/4 Threads EW-56561-01 Mfr # 3-2724-01
COMPARE
pH Electrode, flat surface, Pt-1000 ATC, ISO 7/1 R3/4 threads
$414.25USD / Each
GF Signet 3-2724-10 pH Electrode, Flat Surface, 3 kOhm Balco Thermistor ATC, 3/4" NPT Threads EW-56561-02 Mfr # 3-2724-10
COMPARE
pH Electrode, flat surface, 3KOhm Balco thermistor ATC, 3/4\" NPT threads
$406.00USD / Each
GF Signet 3-2724-11 pH Electrode, Flat Surface, 3 kOhm Balco Thermistor, ISO 7/1 R3/4 Threads EW-56561-03 Mfr # 3-2724-11
COMPARE
pH Electrode, flat surface, 3KOhm Balco thermistor, ISO 7/1 R3/4 threads
$735.00USD / Each
GF Signet 3-2726-HF-00 pH Electrode, HF Resistant, Pt-1000 ATC, 3/4" NPT Threads EW-56561-04 Mfr # 3-2726-HF-00
COMPARE
pH Electrode, bulb, HF resistant, Pt-1000 ATC, 3/4\" NPT threads
$616.00USD / Each
GF Signet 3-2726-HF-10 pH Electrode, HF Resistant, 3 kOhm Balco Thermistor, 3/4" NPT Threads EW-56561-06 Mfr # 3-2726-HF-10
COMPARE
pH Electrode, bulb, HF resistant, 3KOhm Balco thermistor, 3/4\" NPT threads
$529.50USD / Each
GF Signet 3-2726-00 pH Electrode, Pt-1000 ATC, 3/4" NPT Threads
Always in Stock
EW-56561-08 Mfr # 3-2726-00
COMPARE
pH Electrode, bulb, Pt-1000 ATC, 3/4\" NPT threads
$386.25USD / Each
GF Signet 3-2726-10 pH Electrode, 3 kOhm Balco Thermistor, 3/4" NPT Threads EW-56561-10 Mfr # 3-2726-10
COMPARE
pH Electrode, bulb, 3KOhm Balco thermistor, 3/4\" NPT threads
$385.25USD / Each
GF Signet 3-2726-LC-00 pH Electrode, Low Conductivity, Pt-1000 ATC, 3/4" NPT Threads EW-56561-12 Mfr # 3-2726-LC-00
COMPARE
pH Electrode, low conductivity, bulb, Pt-1000 ATC, 3/4\" NPT threads
$446.00USD / Each
GF Signet 3-2726-LC-10 pH Electrode, Low Conductivity, 3kOhm Balco Thermistor, 3/4" NPT Threads EW-56561-14 Mfr # 3-2726-LC-10
COMPARE
pH Electrode, low conductivity, bulb, 3KOhm Balco thermistor, 3/4\" NPT threads
$397.00USD / Each
GF Signet 3-2725-60 ORP Electrode, Flat Surface, 3/4" NPT Threads EW-56561-16 Mfr # 3-2725-60
COMPARE
ORP Electrode, flat surface, 3/4\" NPT threads
$477.50USD / Each
GF Signet 3-2725-61 ORP Electrode, Flat Surface, ISO 7/1 R3/4 Threads EW-56561-17 Mfr # 3-2725-61
COMPARE
ORP Electrode, flat surface, ISO 7/1 R3/4 threads
$530.00USD / Each
Showing 1- 12 of 12