General Tools Digital Manometers
Ideal for laboratory research equipment, computer peripherals, pneumatic controls, HVAC and more
  • Select from one of 5 pressure units of measure: inH2O, psi, mbar, bar, mmH2O
  • Bright backlight
  • Data hold & record features
  • Min/max button
  • Simple fittings for hose connection
  • Auto power off & low battery indicator
Displays gauge or differential pressure.
$273.50 - $315.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
General Tools DM8200 Digtl Manomtr 100PSI 150PSI Max Pressure EW-97008-76 Mfr # DM8200
COMPARE
$285.25USD / Each
General Tools DM8215 Dgtl Manomtr 0/15PSI Standard Digital Manometer 30PSI Max Pressure EW-97008-77 Mfr # DM8215
COMPARE
$273.50USD / Each
General Tools DM8252 Digital Manometer, 0/2PSI, High Resolution EW-97008-79 Mfr # DM8252
COMPARE
$315.00USD / Each