This product is now discontinued, View Related Products
GE Sensing Druck DPI 612 Flex Series Pressure Calibrators
Reduce calibration time with large memory and simple operation
  • Perform calibrations with high accuracy, touchscreen control, and a self-contained pressure test and calibration station
  • Chose the pressure range to meet your application needs—pressure modules are interchangeable
  • Perform "hot swaps" of pressure modules without needing tools, seals, or cables
  • Avoid leaks with quick-to-fit pressure adapters and hoses—no tools or seals required
  • Document test results and save test settings on the 8 Gb internal memory
  • Avoid rework using the PASS/FAIL error analysis feature
  • Electrical measurement and sourcing capability
In the fifth generation of Druck DPI 600 pressure calibrators, the DPI 612 Flex series offers high accuracy, improved generation capabilities, and simple touchscreen operation. Three models cover ranges of 95% vacuum (nominal) up to 300, 1500, or 15,000 psi (20, 100, or 1000 bar) and have an 8 Gb internal memory for data and setting storage. Test settings may also be saved internally, reducing set up and calibration time from 40 minutes to less than 10 minutes!
Simplified touchscreen controls handle much like a smartphone or tablet—the application-like dashboard allows for quick task and feature selection. Move applications and save settings as favorites to obtain task changes in as little as three steps. Advanced features/applications include PASS/FAIL, data logging, software interaction, automated calibration procedures, step and ramp mA output, nudge for incremental mA changes, switch testing, pressure leak testing, MIN/MAX, and adjustable relief valve, scaling, and resolution settings.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GE Druck DPI 612 pFlex Sensing Pressure Calibrator EW-68974-35 Mfr # DPI 612-PFX-00
COMPARE
Sensing Pressure Calibrator
Call For Price
GE Druck DPI 612 pFlex Sensing Pressure Calibrator Kit with -14.5 to 300 psig Module EW-68974-37 Mfr # DPI 612-PFX-20G
COMPARE
Sensing Pressure Calibrator Kit with -14.5 to 300 psig Module
Call For Price
GE Druck DPI 612 pFlexPro Sensing Pressure Calibrator EW-68974-39 Mfr # DPI 612-PFP-00
COMPARE
Sensing Pressure Calibrator
Call For Price
GE Druck DPI 612 pFlexPro Sensing Pressure Calibrator Kit with -14.5 to 300 psig Module EW-68974-41 Mfr # DPI 612-PFP-35G
COMPARE
Sensing Pressure Calibrator Kit with -14.5 to 300 psig Module
Call For Price
GE Druck DPI 612 hFlexPro Sensing Pressure Calibrator EW-68974-43 Mfr # DPI 612-HFP-00
COMPARE
Sensing Pressure Calibrator
Call For Price
GE Druck DPI 612 hFlexPro Sensing Pressure Calibrator Kit with 0 to 10,000 psi Module EW-68974-45 Mfr # DPI 612-HFP-700
COMPARE
Sensing Pressure Calibrator Kit with 0 to 10,000 psi Module