Fuego SCS Safety Enhanced Laboratory Gas Burners
Simple and safe operation for all flame related applications in the laboratory
  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
With an ergonomic design and extensive standard features, these laboratory gas burners withstand extreme lab conditions while incorporating innovative safety technology. The Safety Control System (SCS) constantly analyzes potential hazards, and, if necessary, initiates safety measures, such as an interruption of the gas supply. An adjustable automatic cut-off system inhibits unintentional ignition. To protect you from burns after prolonged use, the residual heat display will signal that the burner head is still hot. Burners also feature continuous burner head control which identifies burner head clogging and ensures correct assembly of burner head.
$958.00 - $1,241.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Gas Type Inlet Tube OD (in) Description
Availability Pricing
Fuego 8.001.000 Scs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane EW-36302-02 Mfr # 8,201,000
COMPARE
Natural Gas, Propane/Butane 0.25 Scs Basic Laboratory Gas Burner; Natural Gas, Propane/Butane
$958.00USD / Each
Fuego SCS Burner with Infrared Sensor EW-36302-04 Mfr # 8,200,000
COMPARE
Natural Gas, Propane/Butane 0.25 Burner with Infrared Sensor
$1,066.00USD / Each
Fuego 8.004.000 Scs Pro -- with Infrared and Rechargeable Battery EW-36302-06 Mfr # 8 204 000
COMPARE
Natural Gas, Propane/Butane 0.25 Scs Pro -- with Infrared and Rechargeable Battery
$1,241.00USD / Each