Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
Fluoroplastic pumps with carbon fiber-reinforced ETFE are designed for highly corrosive or high-purity applications.
$818.25 - $1,638.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Power (VAC)
Availability Pricing
COMPARE
3.2 26 115
$861.75USD / Each
COMPARE
3.2 26 230
$818.25USD / Each
COMPARE
16 26 115
$1,473.50USD / Each
COMPARE
36 38 230
$1,638.50USD / Each