This product is now discontinued, View Related Products

Fluke Waveform Function Generators

Easily generate nonstandard and user-defined waveforms
  • Multiple standard waveforms recalled from internal memory
  • Built-in trigger burst
  • Includes RS-232 and GPIB interfaces
These function generators use direct digital synthesis techniques . Multiple standard and complex waveforms are recalled from internal memory.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Frequency Range (Hz) Number Of Channels Sample Rate
Availability Pricing
Fluke 271-U 115V DDS Function Generator, 10 MHz EW-20018-61 Mfr # 271-U 115V
COMPARE
10000000 1 27.48M samples/sec
Call For Price
Fluke 281-U 115V 40 MS/s Universal Waveform Function Generator EW-20018-62 Mfr # 281-U 115V
COMPARE
16000000 1 40M samples/sec
Call For Price
Fluke 282-U 115V 40 MS/s 2-Channel Waveform Function Generator EW-20018-63 Mfr # 282-U 115V
COMPARE
2 40M samples/sec
Call For Price
Fluke 284-U 115V 40 MS/s 4-Channel Waveform Function Generator EW-20018-64 Mfr # 284-U 115V
COMPARE
4 40M samples/sec
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat