Fluke® T6 Electrical Testers with FieldSense Technology
Measure voltage without leads or clamps
  • Place up to AWG 4/0 wires in the open fork to obtain readings
  • Obtain AC and DC voltage and current measurements
  • T6-1000 also measures frequency and resistance
  • Features backlit display and HOLD button to temporarily freeze the display
These electrical testers feature noncontact FieldSense technology to make your job faster, easier, and safer. The open-fork design accepts up to AWG 4/0 wires (17.8 mm diameter)—no need for leads, clamps, opening covers, or removing wire nuts to obtain TRMS voltage and current readings.
Connect the included test leads to increase accuracy and obtain VDC readings as well.
Measure up to 600 volts with the T6-600 with a CATIII 600 V safety rating—or 1000 volts with the T6-1000 and a CATIII 100 V safety rating. The T6-1000 also displays voltage and current readings simultaneously.
ITEM(S) ON SALE
$224.99 - $332.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) Description
Availability Pricing
Fluke T6-1000 Electrical Tester with FieldSense Technology; 1000 V EW-20004-45 Mfr # T6-1000
COMPARE
$332.99USD / Each
$369.99USD / Each
Fluke T6-600 Electrical Tester with FieldSense Technology; 600 V EW-20004-65 Mfr # T6-600
COMPARE
$224.99USD / Each
$249.99USD / Each