Fluke T-Series Electrical Testers
Troubleshoot problems safely and quickly
  • Available in 600 volt and 1000 volt models
  • OpenJaw™current feature lets you measure current up to 100 amps without breaking the circuit
  • Digital display
  • Rotary switch selects volts, amps and ohms function
  • Detachable SlimReach™test probes reach into tight places, fit snugly into wall sockets, and stow neatly in the back of the testers
  • Optional H5 holster keeps the tester leads neat and lets you clip the T5 onto your belt
  • Rugged design—testers withstand drops from up to 10 feet!
Fluke T5 electrical testers let you check voltage, continuity and current with one compact tool. Troubleshoot a wider range of problems safer and faster than a traditional solenoid tester.
ITEM(S) ON SALE
$153.98 - $255.05USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) AC Max (Volts) Resistance Max (Ω)
Availability Pricing
Fluke T5-1000 Electrical Tester Kit with Holster and 1AC II Voltage Tester EW-20008-38 Mfr # T5-H5-1AC KIT/US
COMPARE
100 1000 1000
$255.05USD / Each
$264.99USD / Each
Fluke T5-600 USA 600V Voltage, Continuity and Current Tester EW-26078-10 Mfr # T5-600 USA
COMPARE
100 600 1000
$153.98USD / Each
$159.99USD / Each
Fluke T5-1000 USA 1000V Voltage and Continuity Tester EW-26078-20 Mfr # T5-1000 USA
COMPARE
100 1000 1000
$192.48USD / Each
$199.99USD / Each