Fluke PRV240 Proving Unit Kits
Validate the functionality of test tools without placing yourself in a potentially hazardous electrical environment
  • “Test before touch”—reduce unnecessary exposure to shock and arc flash hazards
  • Compatible with high or low impedance multimeters, clamp meters, or two pole testers
  • Sources both AC and DC steady-state voltage
Fluke proving unit kits combine the PRV420 proving unit with either a Fluke multimeter, clamp meter, or electrical tester providing you optimal flexibility based on your application requirements. These kits are ideal for electrical maintenance personnel, electrical contractors, and electricians in the electrical, oil and gas, utilities markets, and more.
These kits, with included PRV240 proving unit, are designed for safety and compliance applying the concept of test before touch (TBT) that involves testing your meter against a known live source before and after measurements. This verifies that your test tool is operating properly during the actual measurement without exposing yourself to shock or arc flash.
ITEM(S) ON SALE
$360.92 - $476.99USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Fluke FLK-3000FC/PRV240 Proving Unit Kit with 3000FC Multimeter EW-20049-51 Mfr # FLK-3000FC/PRV240
COMPARE
$476.99USD / Each
$529.99USD / Each
Fluke 117/PRV240 Proving Unit Kit with 117 Multimeter EW-20049-53 Mfr # FLUKE-117/PRV240
COMPARE
$391.49USD / Each
$434.99USD / Each
Fluke T5-1K/PRV240 Proving Unit Kit with T5-1000 Electrical Tester EW-20049-55 Mfr # FLUKE-T5-1K/PRV240
COMPARE
$360.92USD / Each
$374.99USD / Each