Fluke Leakage Current Clamp Meters with Wireless Data Transfer
Accurate True RMS reading of current signals as low as 10 µA
  • Transmit results wirelessly via Fluke Connect® app
  • Selectable filter function removes unwanted noise
  • Forward-facing LED work light brightens dark work areas
This True RMS leakage current clamp meter enables technicians to accurately identify, document, record, and compare leakage current readings over time to prevent potential problems and identify intermittent GFCI and RCD trips, all without taking equipment offline. Measurements can be synced wirelessly to the cloud and referenced any time from your smartphone, tablet, or desktop by using the free Fluke Connect app. The fully shielded, oversized clamp jaw easily accommodates larger conductors and minimizes external EMI to accurately capture leakage current signals as low as 10 µA.
The rugged CAT III-600 V safety rated meter also measures complex signals with the highest resolution of 1 µA and an upper measurement range of 60 A. Features include Min/Max/Average readings and hold function, forward-facing LED worklight designed for poorly lit areas, backlit display, and auto power-off for extended battery life.
Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$845.99 - $1,099.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fluke 368 FC Leakage Current Clamp Meter with Wireless Data Transfer, 1-9/16" (40 mm) dia clamp EW-20006-79 Mfr # FLUKE-368 FC
COMPARE
Leakage Current Clamp Meter with Wireless Data Transfer, 1-9/16\" (40 mm) dia clamp
$845.99USD / Each
$939.99USD / Each
Fluke 369 FC Leakage Current Clamp Meter with Wireless Data Transfer, 2-13/32" (61 mm) dia clamp EW-20006-80 Mfr # FLUKE-369 FC
COMPARE
Leakage Current Clamp Meter with Wireless Data Transfer, 2-13/32\" (61 mm) dia clamp
$1,099.99USD / Each