Fluke Calibration (Hart Scientific) 5627A Series - Precision industrial PRT/RTD probes
5627A Series – Precision industrial PRT/RTD probes: One of the best features of this sensor is that it conforms to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a coil suspension element design for increased shock and vibration resistance. It has a mineral-insulated sheath with a minimum bend radius of 19 mm (3/4-inch) for flexibility and durability.
Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$954.90USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
Fluke Calibration 5627A-12-L High Temperature PRT-100 Ohm 4 Wire EW-10508-71 Mfr # 5627A-12-L
COMPARE
Mini Spade Lug 788
$954.90USD / Each
$1,061.00USD / Each
Fluke Calibration 3003572 Industrial PRT Probe, 12" , Din EW-90450-83 Mfr # 5627A-12-D
COMPARE
5-Pin DIN 788
$954.90USD / Each
$1,061.00USD / Each
Fluke Calibration 3377081 High Accuracy Temperaure PRT Probe, 12" EW-90450-85 Mfr # 5627A-12-P
COMPARE
Info-Con 788
$954.90USD / Each
$1,061.00USD / Each
Fluke Calibration 3003648 Industrial PRT Probe, 6" , B-Wire EW-90450-86 Mfr # 5627A-6-B
COMPARE
Stripped Wire 788
$954.90USD / Each
$1,061.00USD / Each
Fluke Calibration 3377096 Industrial PRT Probe, 6" Info-Con EW-90450-89 Mfr # 5627A-6-P
COMPARE
Info-Con 788
$954.90USD / Each
$1,061.00USD / Each
Fluke Calibration 3377107 Industrial PRT Probe, 9" Info-Con EW-90450-93 Mfr # 5627A-9-P
COMPARE
Info-Con 788
$954.90USD / Each
$1,061.00USD / Each