Fluke Calibration (Hart Scientific) 5623B Series - Full immersion freezer PRT/RTD probes
5623B Series - Full immersion freezer PRT/RTD probes
: Use this probe when calibrating freezers, freeze dryers and other cold chambers. This unique freezer probe measures with ±0.05°C accuracy, even when its entire handle and cable are immersed in temperatures as cold as –100°C. Other PRTs will break when the entire assembly is exposed to the temperatures that this probe can handle.
Available for sale in the United States and Canada only.
$1,424.00 - $1,435.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Fluke Calibration 3003488 PRT/RTD Freezer Probe, 0.25" D x 6" L; Wire EW-90450-62 Mfr # 5623B-6-B
COMPARE
Stripped Wire 313 -148
$1,424.00USD / Each
Fluke Calibration 3377043 PRT/RTD Freezer Probe, 0.25" D x 6" L; I-Con EW-90450-65 Mfr # 5623B-6-P
COMPARE
Info-Con 313 -148
$1,435.00USD / Each