Fluke Calibration (Hart Scientific) 5610 Series - Secondary thermistor probes
5610 Series - Secondary thermistor probes: The small drift and low calibration uncertainty of only ±0.01°C makes this rugged stainless steel thermistor a wise choice when temperatures of interest are between 0 and 100°C. These probes are ideal for measuring temperature in air, liquid or soil.
Available for sale in the United States and Canada only.
$1,336.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Description
Availability Pricing
Fluke Calibration 3376859 Thermistor Probe; 1/8" Dia x 6" Linfo-Con 1523/24 EW-90451-06 Mfr # 5610-6-P
COMPARE
Info-Con 212 Thermistor Probe; 1/8\" Dia x 6\" Linfo-Con 1523/24
$1,336.00USD / Each