Fluke Calibration (formerly Hart Scientific) Tweener High-Accuracy Thermometers
1502 Accuracy of ±0.009°C at 100°C
1504 Accuracy of ±0.004°C at 50°C
  • Built-in RS-232 interface lets you automate data collection, calibration, and process control functions via your computer
Easily program this thermometer using the front panel push-button keypad-programmable to match anyprobe characteristics. View temperature in degF, degC, K, or ohms on the large LED display. Select temperature scale readout according to IPTS-68 or ITS-90. Sampling rate is two readings per second. Battery backup memory saves setup information and probe constants when the power is turned off. Precision probes feature PFE-insulated leads and high-stability sensor to provide accurate readings. Meter and probe systems include a certificate with four calibration points and R0, alpha, delta, A4, and C4 calibration constants.
Available for sale in the United States and Canada only.
$3,738.00 - $3,750.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
Fluke Calibration 1504 Temperature Benchtop Meter Tweener Thermistor EW-90448-92 Mfr # 1504-156
COMPARE
1764 Thermistor 1
$3,738.00USD / Each
Fluke Calibration 1502A-156 Temperature Benchtop Meter Tweener EW-93201-15 Mfr # 1502A-156
COMPARE
RTD 1
$3,750.00USD / Each