Fluke Calibration (formerly Hart Scientific) Deep Well Compact Baths
Hart's deep-well compact baths are small, fast and quiet, yet with enough depth to calibrate even the longest PRTs, SPRTs, and liquid-in-glass (LIG) thermometers.
  • Access opening 6.8 x 4.7 in; 18 in deep, or 19 in with optional fluid level adapter
  • RS232 ports for automated calibrations with optional MET / TEMP II software
  • Fast heat-up and cool-down times
  • Small, 4.2 gal (15.9L) fluid volume with expansion reservoir to catch fluid overflow
  • Optional LIG thermometer calibration kit and magnifying scope
Available for sale in the United States and Canada only.
$2,766.79 - $19,297.43USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Power (VAC)
Availability Pricing
Fluke Calibration 7341-16 Deep Well Compact Bath, -45/50C EW-16101-71 Mfr # 7341-16
COMPARE
302 -49 115
$19,297.43USD / Each
Fluke Calibration 2019-DC8 Fluid Level Adapter Kit EW-16103-32 Mfr # 2019-DC8
COMPARE
115
$2,766.79USD / Each
Fluke Calibration 6331-B Bath, Deep Well, Compact 40 to 300 C EW-39006-61 Mfr # 6331-156
COMPARE
572 104 115
$13,393.00USD / Each