Fluke 721 Precision Pressure Calibrator
Provide pressure calibration and temperature measurement for custody transfer applications
  • Both pressure inputs perform pressure switch tests
  • Large backlit graphic LCD displays up to three inputs simultaneously
  • Store five instrument setups for quick recall of routine tests
  • Choose between standard and intrinsically safe models
Dual-isolated pressure sensors make the Fluke 721 precision calibrator the ideal tool for gas custody transfer applications, allowing you to take simultaneous static and differential pressure measurements with one calibrator.
Standard 721 models (16107-22 through -35) feature an internal 24 V loop power supply to power a transmitter under test, and measures up to 30 VDC to check 24 V loop power supplies. Extend pressure measurement range with connection to external 700P/750P pressure modules.
3 PRODUCT OPTIONS
Compareitem specifications
Max Measurement (VDC) Min Pressure Pressure Resolution
Availability Pricing
Fluke 721-1601 Precision Pressure Calibrator, dual 16 and 100 psig sensors EW-16107-22 Mfr # Fluke-721-1601
COMPARE
24 16 psig or 100 psig 0.01 psi (0.0001 bar) / 0.01 psi (0.0001 bar)
$3,487.99USD / Each
Fluke 721-1650 Precision Pressure Calibrator, dual 16 and 5000 psig sensors EW-16107-28 Mfr # Fluke-721-1650
COMPARE
24 16 psig or 5000 psig 0.01 psi (0.0001 bar) / 0.1 psi (0.01 bar)
$3,487.99USD / Each
Fluke 721-3601 Precision Pressure Calibrator, dual 36 and 100 psig sensors EW-16107-29 Mfr # Fluke-721-3601
COMPARE
24 36 psig or 100 psig 0.01 psi (0.0001 bar) / 0.01 psi (0.0001 bar)
$3,487.99USD / Each