Fluke® 500 Series Battery Analyzers
Intuitive easy-to-use analyzers reduce battery testing complexity
  • Battery resistance, AC/DC voltage and current, ripple voltage, frequency and battery temperature functions allow quick assessment of battery conditions
  • Automatic or manual sequence measurement modes allow the testing of battery strings with automatic measurement storage
  • Unit automatically captures all measured values during testing, providing on the-go analysis capability
  • Intuitive user interface includes visual and audio feedback
  • Threshold comparison function allows you to configure multiple reference values for resistance and voltage
  • Enhanced data analysis capability allows you to quickly compare trends, analyze results, and create reports with included battery management software
These Fluke 500 series battery analyzers are designed for maintenance, troubleshooting and performance testing of individual stationary batteries and battery banks used in critical battery back-up applications. Analyzers provide high performance, high accuracy, reliable test results to help ensure your batteries are in top operating condition. Units feature a wide range of battery test functions from DC voltage and resistance tests to full condition testing using an automated string function test, along with an integrated infra-red test probe temperature measurement system.
The intuitive user-interface makes these analyzers easy to set up and operate, and the compact rugged design withstands challenging environments. The included PC software allows you to quickly generate PDF reports that include analysis graphics and data tables, or email formats (model BT521 and BT521ANG) with the csv file on the mobile app. Models BT521 and BT521ANG also feature wireless communication.
$6,852.99 - $7,689.99USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Battery Resistance Accuracy DC Max (Volts) Description
Availability Pricing
Fluke BT521ANG Advanced Battery Analyzer with Angled Tip Test Probes EW-20037-50 Mfr # FLUKE-BT521ANG
COMPARE
1% + 68 at 3 mΩ, 0.8% + 12 at 30 mΩ, 0.8% + 6 at 300 and 3000 mΩ 1000 Advanced Battery Analyzer with Angled Tip Test Probes
$7,689.99USD / Each
Fluke BT520ANG Standard Battery Analyzer with Angled Tip Test Probes EW-20037-51 Mfr # FLUKE-BT520ANG
COMPARE
1% + 68 at 3 mΩ, 0.8% + 12 at 30 mΩ, 0.8% + 6 at 300 and 3000 mΩ 600 Standard Battery Analyzer with Angled Tip Test Probes
$6,987.99USD / Each
Fluke BT521 Advanced Battery Analyzer EW-20048-09 Mfr # FLUKE-BT521
COMPARE
1% + 8 at 3 mΩ, 0.8% + 6 at 30, 300, and 3000 mΩ 1000 Advanced Battery Analyzer
$6,852.99USD / Each