Fluke® 370 FC Series True RMS Wireless AC/DC Clamp Meters
Innovative, versatile, and wireless diagnostic tools with increased performance and flexibility
  • Log and trend measurements, and pinpoint intermittent faults
  • Capture measurements outside the arc flash zone with Bluetooth® connectivity to your Apple or Android devices
  • Transmit results wirelessly via the Fluke Connect® Measurements app
  • Read measurements on your phone at a safe distance, requiring less Personal Protective Equipment (PPE)
  • Create and share reports from the field via email, or converse in real-time with ShareLive™ video calls
  • CAT IV-600V, CAT III-1000 V safety rating
Fluke® 370 FC wireless series clamp meters offer True RMS AC voltage and current measurements. Depending on model, the meters can measure up to 1000 V and 1000 A or 600 V and 600 A. The iFLEX® flexible current probe (included with 376 FC, must be ordered separately for 374 FC and 375 FC) expands the AC current measurement range to 2500 A while providing increased display flexibility, and the ability to measure awkward-sized conductors and access wires. All three clamps are now part of the Fluke Connect® family of wireless test tools, allowing users to automatically transmit data to their phone.
ITEM(S) ON SALE
$476.99 - $715.49USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) Resistance Max (Ω) Description
Availability Pricing
Fluke IB376K 376 FC True-rms Clamp Meter Plus Insulated Hand Tools Starter Kit EW-20036-49 Mfr # IB376K
COMPARE
1000 60000 IB376K 376 FC True-rms Clamp Meter Plus Insulated Hand Tools Starter Kit
$715.49USD / Each
$794.99USD / Each
Fluke 374 FC 600 A AC/DC True-rms Wireless Clamp Meter EW-20050-89 Mfr # FLUKE-374 FC
COMPARE
600 6000 374 FC 600 A AC/DC True-rms Wireless Clamp Meter
$476.99USD / Each
$529.99USD / Each
Fluke 375 FC 600 A AC/DC True-rms Wireless Clamp Meter EW-20050-90 Mfr # FLUKE-375 FC
COMPARE
600 60000 375 FC 600 A AC/DC True-rms Wireless Clamp Meter
$521.99USD / Each
$579.99USD / Each
Fluke 376 FC 1000 A AC/DC True-rms Wireless Clamp Meter with iFLEX® 2500 A Probe EW-20050-91 Mfr # FLUKE-376 FC
COMPARE
1000 60000 376 FC 1000 A AC/DC True-rms Wireless Clamp Meter with iFLEX® 2500 A Probe
$593.99USD / Each
$659.99USD / Each