Fluke 323, 324, and 325 400 ATrue RMS Clamp Meters
Precision troubleshooting clamp meters ideal for commercial and residential applications
  • True RMS AC voltage and current for accurate measurements on nonlinear signals
  • Data hold function for additional measurement flexibility
  • Slim, ergonomic, rugged design
The 323, 324 and 325 are designed to verify the presence of load current, AC voltage and continuity of circuits, switches, fuses and contacts.
3 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) Resistance Max (Ω) Description
Availability Pricing
Fluke 323 TRMS Clamp Meter, 400 A, with resistance EW-20043-74 Mfr # FLUKE-323
COMPARE
600 4000 TRMS Clamp Meter, 400 A, with resistance
$179.99USD / Each
$199.99USD / Each
Fluke 324 TRMS Clamp Meter, 400 A, with resistance and capacitance EW-20043-75 Mfr # FLUKE-324
COMPARE
600 4000 TRMS Clamp Meter, 400 A, with resistance and capacitance
$269.99USD / Each
$299.99USD / Each
Fluke 325 TRMS Clamp Meter, 400 A, with resistance, capacitance, and frequency EW-20043-76 Mfr # FLUKE-325
COMPARE
600 4000 TRMS Clamp Meter, 400 A, with resistance, capacitance, and frequency
$377.99USD / Each
$419.99USD / Each