FLIR® T800 Series High-Performance Thermal Imaging Cameras
Scope out problems before they become costly
  • Comfortably monitor indoor/outdoor equipment with 180° rotating lens
  • Diagnose hard-to-reach components with advanced resolution/image clarity
  • Scan outdoor equipment from a safe distance using integrated eyepiece viewfinder
  • Easily identify the complexity of repairs via on-screen distance measurements
  • Share lenses with other FLIR cameras featuring AutoCal™ optics
  • Instantly convey critical findings to stakeholders using FLIR Tools® mobile app
With image clarity enhanced by FLIR Vision Processing™, MSX®, UltraMax® and proprietary adaptive filtering, the high-performance FLIR T800 series thermal imaging cameras can safely survey equipment for signs of failure from a safe distance. An on-board inspection routing feature speeds data collection and reporting to help you plan out surveys and organized images and data by location.
Laser-assisted autofocus and advanced measurement tools enable accurate temperature readings, while the integrated eyepiece viewfinder and bright, 4" color LCD makes daylight inspections a breeze. Intuitive touchscreen controls simplify reporting, with the capacity to embed GPS locations allowing you to observe critical findings through Wi-Fi streaming to the FLIR Tools® app.
$31,289.00 - $34,659.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Detector Size Max Temperature (° F) Description
Availability Pricing
Flir High-Performance Thermal Imaging Camera, MSX/14 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-66 Mfr # T865-14
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/14 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$31,289.00USD / Each
Flir High-Performance Thermal Imaging Camera, MSX/24 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-67 Mfr # T865-24
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/24 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$31,289.00USD / Each
Flir High-Performance Thermal Imaging Camera, MSX/42 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-68 Mfr # T865-42
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/42 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$31,289.00USD / Each
Flir High-Performance Thermal Imaging Camera, MSX/24 & 14 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-69 Mfr # T865-24-14
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/24 & 14 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$32,979.00USD / Each
Flir High-Performance Thermal Imaging Camera, MSX/24 & 42 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-70 Mfr # T865-24-42
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/24 & 42 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$32,979.00USD / Each
Flir High-Performance Thermal Imaging Camera, MSX/24, 14 & 42 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-71 Mfr # T865-24-14-42
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/24, 14 & 42 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$34,659.00USD / Each
Flir High-Performance Thermal Imaging Camera, MSX/42 & 14 Degree Lens, 640 x 480 Resolution with FLIR Tools+ EW-39757-72 Mfr # T865-42-14
COMPARE
640 x 480 3632 High-Performance Thermal Imaging Camera, MSX/42 & 14 Degree Lens, 640 x 480 Resolution with FLIR Tools+
$32,979.00USD / Each
Showing 1- 7 of 7