Flir CM4X-Series Professional True RMS Clamp Meters
Obtain more accurate readings on smaller-gauged wires
  • Accu-Tip™ technology allows for precise measurements—even on thin wires
  • Easily see readings even in dark areas—features large-digit, backlit display
  • Slim design is rugged and shaped to fit comfortably in hand for years of use
  • Open the jaws to accept a maximum 1-3/16" (30 mm) diameter conductor
Take amperage measurements on smaller-gauged wires more accurately—to a tenth of a digit—with the new CM4X family of clamp meters. Each meter features VFD mode, non-contact voltage detection, MIN/MAX/AVE recording, frequency and diode measurement, data hold, low pass filter for voltage measurement, zero function, and TRMS readings.
The over-molded, thin-form design is rugged enough to withstand a 6.5-ft (2 m) drop but compact enough to fit well in any toolbox—perfect for commercial and residential electrician use. The large-digit, digital display is backlit and easy to read even in low-light conditions.
ITEM(S) ON SALE
$232.04USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) AC Max (Volts) Description
Availability Pricing
Flir CM46 True RMS AC/DC Digital Clamp Meter with Type K; 600V, 400A EW-26848-15 Mfr # CM46
COMPARE
400 600 600V 400A AC/DC Digital TRMS Clamp Meter with Type K
$232.04USD / Each
$272.99USD / Each