This product is now discontinued, View Related Products
Flir A3xxf Series Fix Mount Thermal Cameras with IP66 Enclosure
Accurate temperature readings at an affordable price
  • Housing meets IP-66 requirements
  • Produces 320 x 240 pixel IR resolution thermal images
  • MPEG-4 streamed video
  • C-video
The FLIR fix mount thermal cameras offer an affordable and accurate temperature measurement solution for anyone who needs to solve problems that do not call for the highest speed or reaction, and who uses a PC. Due to its composite video output, it is an excellent choice for thermal image automation applications, where you can utilize its unique properties such as looking through steam.
Package contents include:
• Infrared camera with lens
• Calibration certificate
• Cardboard box
• Downloads brochure
• Ethernet cable
• FLIR apps card
• FLIR Tools download card
• Getting Started Guide
• Important Information Guide
• Mains cable
• Optics brochure
• Power cable, pig-tailed
• Power supply
• Service and training brochure
• Thank you card
• User documentation CD-ROM
• Utility CD-ROM
• Registration card
Automation thermal imagers in many cases require application support prior to ordering. Please contact our technical application group or sales team for support on this product. In some areas of the country onsite field assessments can be arranged.
Call For Price
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Camera Included
Availability Pricing
Flir 61201-1003 Fixed Automation Thermal Camera IP-66 Rated with 25 Deg Lens EW-39755-33 Mfr # FLIR A300F W/25 LENS
COMPARE
A300f 25 Fixed IP-66 Rated with 25 deg Lens
Call For Price
Flir 61201-1004 Fixed Automation Thermal Camera IP-66 Rated with 45 Deg Lens EW-39755-34 Mfr # FLIR A300F W/45 LENS
COMPARE
A300f 45 Fixed IP-66 Rated with 45 deg Lens
Call For Price
Flir 61201-1005 Fixed Automation Thermal Camera IP-66 Rated with 90 Deg Lens EW-39755-35 Mfr # FLIR A300F W/90 LENS
COMPARE
A300f 90 Fixed IP-66 Rated with 90 deg Lens
Call For Price
Flir 61201-1002 Fixed Automation Thermal Camera IP-66 Rated with 15 Deg Lens EW-39755-36 Mfr # FLIR A300F W/15 LENS
COMPARE
A300f 15 Fixed IP-66 Rated with 15 deg Lens
Call For Price
Flir 61201-1001 Fixed Automation Thermal Camera IP-66 Rated with 6 Deg Lens EW-39755-37 Mfr # FLIR A300F W/6 LENS
COMPARE
A300f 6 Fixed IP-66 Rated with 6 deg Lens
Call For Price
Flir 61201-1103 Fixed Automation Thermal Camera IP-66 Rated with 25 Deg Lens EW-39755-38 Mfr # FLIR A310F W/25 LENS
COMPARE
A310f 25 Fixed IP-66 Rated with 25 deg Lens
Call For Price
Flir 61201-1104 Fixed Automation Thermal Camera IP-66 Rated with 45 Deg Lens EW-39755-39 Mfr # FLIR A310F W/45 LENS
COMPARE
A310f 45 Fixed IP-66 Rated with 45 deg Lens
Call For Price
Flir 61201-1105 Fixed Automation Thermal Camera IP-66 Rated with 90 Deg Lens EW-39755-41 Mfr # FLIR A310F W/90 LENS
COMPARE
A310f 90 Fixed IP-66 Rated with 90 deg Lens
Call For Price
Flir 61201-1102 Fixed Automation Thermal Camera IP-66 Rated with 15 Deg Lens EW-39755-42 Mfr # FLIR A310F W/15 LENS
COMPARE
A310f 15 Fixed IP-66 Rated with 15 deg Lens
Call For Price
Flir 61201-1101 Fixed Automation Thermal Camera IP-66 Rated with 6 Deg Lens EW-39755-43 Mfr # FLIR A310F W/6 LENS
COMPARE
A310f 6 Fixed IP-66 Rated with 6 deg Lens
Call For Price
Flir 61201-1203 Fixed Automation Thermal Camera IP-66 Rated and 25 Deg Lens EW-39755-44 Mfr # FLIR A315F W/25 LENS
COMPARE
A315f 25 Fixed IP-66 Rated and 25 deg Lens
Call For Price
Flir 61201-1204 Fixed Automation Thermal Camera IP-66 Rated and 45 Deg Lens EW-39755-45 Mfr # FLIR A315F W/45 LENS
COMPARE
A315f 45 Fixed IP-66 Rated and 45 deg Lens
Call For Price
Flir 61201-1205 Fixed Automation Thermal Camera IP-66 Rated and 90 Deg Lens EW-39755-46 Mfr # FLIR A315F W/90 LENS
COMPARE
A315f 90 Fixed IP-66 Rated and 90 deg Lens
Call For Price
Flir 61201-1202 Fixed Automation Thermal Camera IP-66 Rated and 15 Deg Lens EW-39755-47 Mfr # FLIR A315F W/15 LENS
COMPARE
A315f 15 Fixed IP-66 Rated and 15 deg Lens
Call For Price
Flir 61201-1201 Fixed Automation Thermal Camera IP-66 Rated and 6 Deg Lens EW-39755-48 Mfr # 61201-1201
COMPARE
A315f 6 Fixed IP-66 Rated and 6 deg Lens
Call For Price
Showing 1- 15 of 15