Sherwood Scientific Filter Bags for Non-Sealed Tubs
Safely retains stray particles
  • Allows passage of exit gases
Bags retain stray particles of the sample being fluidized in the tub while allowing the passage of exit gases. Use one large bag for a single two- or five-liter tub assembly; use four small bags for the multi-tub assemblies.
$93.40 - $392.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sherwood Scientific 50035407 Filter Bag for Non-Sealed Tubs, Large, Nomex EW-02658-81 Mfr # 50035407
COMPARE
Filter bag for non-sealed tubs, Large, Nomex
$392.00USD / Each
Sherwood Scientific 50035400 Filter bag for non-sealed tubs, Large, Nylon EW-02658-82 Mfr # 50035400
COMPARE
Filter bag for non-sealed tubs, Large, Nylon
$270.00USD / Each
Sherwood Scientific 50035404 Filter Bag for Non-Sealed Tubs, Large, Polypropylene EW-02658-83 Mfr # 50035404
COMPARE
Filter bag for non-sealed tubs, Large, polypropylene
$132.00USD / Each
Sherwood Scientific 50035402 Filter Bag for Non-Sealed Tubs, Large, Terylene EW-02658-84 Mfr # 50035402
COMPARE
Filter bag for non-sealed tubs, Large, terylene
$160.00USD / Each
Sherwood Scientific 50035401 Filter bag for non-sealed tubs, Small, Nylon EW-02658-88 Mfr # 50035401
COMPARE
Filter bag for non-sealed tubs, Small, Nylon
$93.40USD / Each