This product is now discontinued, View Related Products
External Thread Smooth Skirt Polypropylene Cryogenic Vials
Color-coded caps enable quick sample identification
Vials are free standing with a conical bottom and feature 0.5 ml graduations. Designed for use in the vapor phase of liquid nitrogen and are ideal for storage or transport of material at extremely low temperatures. External threads provide a smooth inner surface. The screw cap incorporates a molded-in sealing ring for a tight, leakproof seal.
Smooth Skirt Cryogenic Vials have printed graduations and smooth inner walls for easy filling and sample preparation. Printed writing area on side for permanent coding. Vials are molded from polypropylene and are freezable to -70°C. Sterilized by gamma radiation for immediate use.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Cap color Tube Bottom Shape Volume (mL)
Availability Pricing
Qorpak AKM-3100-0055 External Thread PP Cryogenic Vials, Sterile/Smooth/1.5 mL; 2000/Cs EW-03756-92 Mfr # AKM-3100-0055
COMPARE
Red Slotted skirt 1.5
Call For Price
COMPARE
White Slotted skirt 5