Extech MA3000-Series 3000A, TRMS Flexible Clamp Meters
Flexible clamp fits any current source and compact size fits any toolbox
  • Small-diameter clamp cable and twist-lock feature enhance versatility
  • True RMS for accurate readings of nonlinear waveforms
  • Autoranging function increases speed and efficiency
  • Backlit LCD is easy to read in any lighting condition
  • CAT III-1000V, CAT IV-600V safety rating
Typically, flexible current clamps are detachable probes for rigid current clamp meters and are designed with long wires connecting clamp to meter. The MA3000-series current meters incorporate only the flexible clamp, foregoing the rigid clamp. Without the extra bulk of a rigid clamp or a long wire connection, this meter is compact and easy to transport in the field. Additional features include overrange and low-battery indicators, and auto power-off with disable function.
$220.50 - $225.75USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Max (amps) Description
Availability Pricing
Extech MA3110 Flexible Clamp Meter, 3000 A AC/DC; 11" Cable EW-20000-76 Mfr # MA3110
COMPARE
±1% 3000 Flexible Clamp Meter, 3000 AAC; 11\" Cable
$220.50USD / Each
Extech MA3018 Flexible Clamp Meter, 3000 AAC; 18" Cable EW-20006-95 Mfr # MA3018
COMPARE
±3% + 5 3000 Flexible Clamp Meter, 3000 AAC; 18\" Cable
$225.75USD / Each