Extech Humidity and Temperature USB Datalogger
Logs humidity and temperature readings with a user programmable sample rate
  • 32,000 readings (16,000 for each parameter: Humidity/Temperature)
  • Dew point indication via Windows® software (included)
  • Selectable data sampling rate: 2s, 5s, 10s, 30s, 1m, 5m, 10m, 30m, 1hr, 2hr, 3hr, 6hr, 12hr, 24hr
  • User-programmable alarm thresholds for RH and Temperature
  • Status Indication via Red/Yellow LED and Green LED
  • Long battery life
ITEM(S) ON SALE
$80.74USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Humidity Accuracy Input Type Max Temperature (° F)
Availability Pricing
Extech RHT10 Temperature and Humidity USB Data Logger EW-26045-45 Mfr # RHT10
COMPARE
±3%RH Temperature and humidity 158
$80.74USD / Each
$94.99USD / Each