Extech High-Intensity Dual-Scale Light Meter
Selection feature compensates for five lighting types
Three-range meter indicates light intensity up to 50,000 lux or 5000 footcandles for five selectable lighting types. The RS-232 interface and optional 4000-reading datalogger module facilitate data documentation (sold separately).
$212.99 - $346.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Certifications
Availability Pricing
Extech 407026 High-Intensity, Dual-Scale Light Meter EW-01588-32 Mfr # 407026
COMPARE
None
$212.99USD / Each
Extech 407026-NIST High-Intensity Dual-Scale Light Meter with NIST-Traceable Calibration Certificate EW-01588-34 Mfr # 407026-NIST
COMPARE
NIST-Traceable
$346.99USD / Each