Extech High Definition VideoScope Inspection Cameras
High definition 5.7" LCD monitor provides premium image quality
  • Mini waterproof (IP57) camera head for high-resolution viewing
  • Four bright LED lamps with dimmer to illuminate viewed object
  • Glare-free close-up field of view
  • SD memory card included to store up to 14,600 images
  • Video recording up to 4 hours with voice annotation
  • AV output for viewing images and video directly on a monitor
  • Video and images can be transferred to your PC via the SD card or USB output
The Extech high definition VideoScope inspection cameras are designed for inspection of small, hard to reach areas. Compact, high-resolution water-proof camera probe and four built-in bright LED lamps with dimmer control illuminate viewed objects and provide glare-free close-up field of view. These videoscopes feature a 5.7” color LCD monitor with high definition 640 x 480 resolution to provide a clear, premium quality video.
$2,099.00 - $3,829.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Lens Shaft Diameter (mm) Description
Availability Pricing
Extech High Definition Videoscope Inspection Camera, 5.5 mm Flexible Probe EW-68517-13 Mfr # HDV610
COMPARE
Attached 5.5 mm Flexible Probe 5.5 High Definition Videoscope Inspection Camera, 5.5 mm Flexible Probe
$2,099.00USD / Each
Extech High Definition Videoscope Inspection Camera, 5.8 mm Flexible Probe EW-68517-15 Mfr # HDV620
COMPARE
Attached 5.8 mm Flexible Probe 5.8 High Definition Videoscope Inspection Camera, 5.8 mm Flexible Probe
$2,099.00USD / Each
Extech High Definition Articulating Videoscope Inspection Camera, 6 mm Flexible Probe EW-68517-17 Mfr # HDV640
COMPARE
Attached Articulating 6 mm Flexible Probe 6 High Definition Articulating Videoscope Inspection Camera, 6 mm Flexible Probe
$3,299.00USD / Each
Extech High Definition Articulating Videoscope Inspection Camera, Wireless, 6 mm Flexible Probe EW-68517-19 Mfr # HDV640W
COMPARE
Wireless Articulating 6 mm Flexible Probe 6 High Definition Articulating Videoscope Inspection Camera, Wireless, 6 mm Flexible Probe
$3,829.00USD / Each