Extech Economical DC Power Supplies
Solid state units provide a constant power supply
  • Light compact design saves valuable benchspace
  • Supplies up to three amps DC current output
These economical DC power supplies are ideal for bench testing, field service, R & D, educational, and telecommunication applications. Units provide short circuit and overload protection and display both voltage and current on the dual backlit display. Additional features include adjustable constant voltage or constant current output, positive and negative banana plug output jacks, and an illuminated on/off switch.
ITEM(S) ON SALE
$154.69USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
Extech 382200 Single Output Economical DC Power Supply, 30V/1A EW-26849-75 Mfr # 382200
COMPARE
0 to 1 A 0 to 30 VDC < 0.5 mVrms; < 3 mArms
$154.69USD / Each
$181.99USD / Each
Extech 382202 Single Output Economical DC Power Supply, 18V/3A EW-26849-80 Mfr # 382202
COMPARE
0 to 3 A 0 to 18 VDC < 0.5 mVrms; < 3 mArms
$154.69USD / Each
$181.99USD / Each