Extech DO600 Dissolved Oxygen Pocket Testers
Measures in percent saturation or concentration
  • Adjustable altitude and salinity compensation
Features include data hold, auto power off, low-battery indicator, and analog bar graph that indicates trends. Order optional extension cable with probe weight to keep probe submerged and protected.
Meter includes: DO electrode, protective sensor cap, spare membrane cap, electrolyte, neckstrap, and four 1.5 V batteries.
Meter kit adds: 6 replacement membranes with caps, 16-ft (5-m) extension cable, probe weight guard, and hard carrying case.
$276.99 - $393.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech DO600 Dissolved Oxygen Pocket Tester
Always in Stock
EW-53026-20 Mfr # DO600
COMPARE
Dissolved Oxygen Pocket Tester
$276.99USD / Each
Extech DO600-K DO600 Dissolved Oxygen Pocket Tester Kit EW-53026-21 Mfr # DO600-K
COMPARE
DO600 Dissolved Oxygen Pocket Tester Kit
$393.99USD / Each