Extech® Compact Indoor Humidity and Temperature Meter
Simultaneously displays temperature and humidity
  • Built-in memory stores max/min readings
  • Clear button resets max/min measurements
  • Measures humidity between 10% to 99%
  • Low battery indication
These compact thermohygrometers are ideal for monitoring temperature and humidity conditions in enclosed or open spaces. The meters simultaneously display current heat and moisture levels as well as record high and low extremes during the day. Additional features include memory clear function, low-battery indication, and built-in stand.
ITEM(S) ON SALE
$26.17USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Description
Availability Pricing
Extech RHM16 Compact Indoor Temperature and Humidity Meter (Celsius) EW-39758-11 Mfr # RHM16
COMPARE
1 LR44 (included) 99 Compact Indoor Temperature and Humidity Meter (Celsius)
$26.17USD / Each
$30.79USD / Each
Extech RHM15 Compact Indoor Temperature and Humidity Meter (Fahrenheit) EW-39759-08 Mfr # RHM15
COMPARE
1 LR44 (included) 99 Compact Indoor Temperature and Humidity Meter (Fahrenheit)
$26.17USD / Each
$30.79USD / Each