Extech Combination Optical/Laser and Contact Tachometers
Contact and optical modes without adapters
  • Display flips when changing from optical to contact mode
These tachometers feature both contact and optical measurement capabilities. Memory button holds last reading up to 5 minutes and recalls minimum and maximum readings. Display flips with the unit to help identify measuring mode. Model 87700-10 features a laser sighting for spotting targets up to 6 1/2 feet (1.9 m) away.
$303.99 - $354.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Measuring Distance (in) Max RPM Range (rpm) Memory
Availability Pricing
Extech Combination Optical/Contact Tachometer EW-87700-07 Mfr # 461895
COMPARE
12 99999 Last reading, min/max
$303.99USD / Each
Extech Combination Laser/Contact Tachometer EW-87700-10 Mfr # 461995
COMPARE
79 99999 Last reading, min/max
$354.99USD / Each