This product is now discontinued, View Related Products
Extech CA3000 Flex Clamp-On Adapters
Expand your measurement capabilities without investing in a new multimeter
  • Banana plugs allow use with most multimeters in the market
  • Small diameter clamp cable and twist-lock feature enhance versatility
  • Measures AC current in three ranges maximizing resolution
Probes are compatible with any multimeter having standard shrouded banana plug sockets and equivalent CAT safety ratings. Battery capacity indicator also included.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Extech CA3010 Flexible Current Clamp-On Adapter, 3000 A AC, 10" cable EW-20006-82 Mfr # CA3010
COMPARE
Call For Price
Extech CA3018 Flexible Current Clamp-On Adapter, 3000 A AC, 18" cable EW-20006-83 Mfr # CA3018