Extech® Big Digit Thermohygrometers
1 inch Digits on super large LCD
  • Max/Min with ''reset'' function
  • RH accuracy: ±4%RH
  • Temperature accuracy: ±1.8°F, (±1°C)
Designed to monitor relative humidity, and temperature. Features include a super large display, C/F switchable, Min/Max with reset function, low battery indication and a built-in stand for sitting on a desk and hardware for mounting on a wall.
ITEM(S) ON SALE
$39.94 - $53.54USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech Big Digit Thermohygrometer
Always in Stock
EW-37802-10 Mfr # 445703
COMPARE
Big Digit Thermohygrometer
$39.94USD / Each
$46.99USD / Each
Extech Big Digit Thermohygrometer with Dew Point, Alarm, and Remote Probe EW-37803-26 Mfr # 445815
COMPARE
Big Digit Thermohygrometer with Dew Point, Alarm, and Remote Probe
$53.54USD / Each
$62.99USD / Each
Extech Big Digit Thermohygrometer with Remote Temperature Probe EW-37809-15 Mfr # 445713
COMPARE
Big Digit Thermohygrometer with Remote Temperature Probe
$45.80USD / Each
$53.89USD / Each
Extech Big Digit Thermohygrometer with Remote Probe EW-37809-16 Mfr # 445715
COMPARE
Big Digit Thermohygrometer with Remote Probe
$50.99USD / Each
$59.99USD / Each
Extech Big Digit Thermohygrometer with Dew Point and Alarm EW-37809-17 Mfr # 445814
COMPARE
Big Digit Thermohygrometer with Dew Point and Alarm
$45.04USD / Each
$52.99USD / Each