Extech 400A Dual Input Clamp Meters With IR Thermometer and NCV
Built-in dual thermocouple inputs for additional flexibility
  • Clamp Meters Feature:
  • True RMS measurements for accurate AC Voltage and Current measurements
  • Dual type K thermocouple input with Differential Temperature function (T1, T2, T1-T2)
  • Built-in non-contact Voltage detector with LED alert
  • DC ìA multimeter function for HVAC flame rod Current measurements
  • Data Hold plus fast Peak Hold of current surges during motor startup
  • 40,000 count multimeter functions for DC Voltage, Resistance, Capacitance, and Frequency for high resolution over wider ranges
  • Measures motor capacitors to 40,000ìF
  • 1.4" (36mm) jaw opening for conductors up to 500MCM
  • Rugged double molded housing
  • Auto power off with audible alert and disable function
  • Autoranging with manual override
  • EX622 and EX623 Additional Features:
  • Built-in non-contact
$266.99USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) Description
Availability Pricing
Extech EX623 400A True RMS, AC/DC, Dual Input Clamp Meter + IR Thermometer EW-26823-46 Mfr # EX623
COMPARE
600 600 400A True RMS, AC/DC, Dual Input Clamp Meter + IR Thermometer
$266.99USD / Each