Eurotherm Digital Temperature 3200-Series Controllers
  • FEATURES
  • Heater current monitoring and load diagnostics
  • Timer functions
  • 8 segment programmer
  • MODBUS communications
  • Setpoint programmer
  • DC control or retrains
Available in three standard DIN sizes the innovative 3200 controllers provide precise temperature control with a host of options. The front panel ‘ammeter’ on the 1/8 and 1/4 DIN sizes displays the heater current or the output power demand. The emphasis is simplicity. A simple ‘Quick Start’ code is used to configure all of the functions essential to controlling your process. This includes the input sensor type, temperature range, heating and cooling outputs, and alarms. In operation every parameter is accompanied by a scrolling text message describing its function. This is available in English, German, French, Spanish or Italian. Custom event and alarm messages can be downloaded from a PC.
$417.00 - $821.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
DIN Size Power (VAC)
Availability Pricing
Eurotherm Temperature Controller, 1/6-DIN 2 Relay Output 1, Output AC Supply EW-90440-21 Mfr # 3216/CC/VH/LR/R/XXX/G/ENG/ENG/XXX/XXXXX/XXXXX
COMPARE
1/16 100 to 240
$417.00USD / Each
Eurotherm Temperature Controller, 1/8-DIN; 2 Relay/4-20mA/Logic/RS-485 EW-90440-24 Mfr # 3208/CC/VH/LRD/R/4CL/G/ENG/ENG/XX/XXXXX
COMPARE
1/8 100 to 240
$821.00USD / Each
Eurotherm Temperature Controller, 1/16-DIN 2 Relay Output 1, 4/20MA Output AC Supply EW-90440-26 Mfr # 3216/CC/VH/DR//R/XXX/G/ENG/ENG/XXX/XXXXX/XXXXX
COMPARE
1/16 100 to 240
$444.00USD / Each