Eurotherm Digital Temperature 2400-Series Controllers
Highly stable with advanced programmable features.
The 2400 series are high stability controllers with an extensive range of options. Either PID, on/off or motorized valve control can be configured, satisfying both electrical and gas heating applications. Dual PID settings, advanced ‘one shot’ and continuously adaptive tuning are available to optimize control performance. These controllers employ universal inputs with Instant Accuracy™ and accept a range of plug-in modules for heating, cooling, analog retransmission, second process value input and remote setpoint. High speed Modbus® RTU, Profibus® DP, DeviceNet™, and Ei-Bisynch ASCII digital communications provide easy connection to PLC™, PC based supervisory control and data logging systems. Multi-zone programming can be implemented using PDSIO® retransmission to deliver a master setpoint to one, two, or three client controllers, with holdback from any client if the temperature deviates from the setpoint by more than a set value.
$206.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
Eurotherm COMMS BOARDMBUS EIA422 EW-94470-33 Mfr # SUB24/EIA422
COMPARE
$206.50USD / Each