Erlab Halo 32 1C Laboratory Grade Air Purification Stations
Provides healthier, cleaner air in your laboratory
  • Healthier air with considerable savings
  • Simple to install and use
  • Easy access panel for quick filter changes
  • BACnet capable
  • Network the units to monitor remotely
Halo 32 1C Laboratory Grade Air Purification Station is easily installed in the ceiling, but does not need to be tied in to existing HVAC. As a standalone unit, it is designed to effectively clean 250-300 sq. ft of space, and it can be placed anywhere that would be most effective for you, You can use multiple units to cover larger areas. It works to purify the air from fugitive emissions present in the lab, while also providing real-time communication and 24/7 data with the lab manager and users of the air pollution levels.
Choose from two filter options using semiconductor sensors for detection and Neutrodyne carbon filters for purification. Chemplus provides broad-spectrum protection while the VOC filter is specific to volatile organic carbon compounds. In addition to purifying the lab air, the Halo is also effective in the reduction of the air exchange rates per hour (ACH) needed to reduce possible concentration of pollutants. Comes with SMART technology for detection and communication capabilities.
$3,705.00 - $4,090.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exhaust Volume (CFM) Filter Type Power (VAC)
Availability Pricing
Erlab Halo 32 1C Chemplus Laboratory Grade Air Purification Station; 80 to 240 VAC EW-33520-31 Mfr # HALOCHEMPLUS
COMPARE
135 Solvents and acids 85 to 240
$4,090.00USD / Each
Erlab Halo 32 1C VOC Laboratory Grade Air Purification Station; 80 to 240 VAC EW-33520-33 Mfr # HALOSMARTVOC
COMPARE
135 Volatile organic compounds 85 to 240
$3,705.00USD / Each