Environmental Express® Universal Solids Standards (USS)
Create accurate and inexpensive control samples
  • Obtain batch level quality analysis and control for all solids testing at an affordable price
  • Quality guaranteed—NIST-traceability is shown on the Certificate of Analysis and each parameter has an acceptability range of ±3 standard deviations
  • Choose from three ranges to best fit your testing needs
These Environmental Express universal solids standards cover multiple EPA test method standard control requirements including TS, TSS, VS, and VSS. Use is easy—simply add the bottle contents into your graduated cylinder and fill to volume, or use as a Matrix Spike by adding a bottle directly to your sample.
$74.70 - $88.00USD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Universal Solids Standard, Low Range; 10/Pk
$76.50USD / Pkg of 10
COMPARE
Universal Solids Standard, High Range; 10/Pk
$74.70USD / Pkg of 10
COMPARE
Universal Solids Standard, Ultra-High Range; 10/Pk
$88.00USD / Pkg of 10