Environmental Express® Kjeldahl Tubes
Optimize your TKN analysis for better results
  • Interchangeable glassware works with SimpleDist and TKN HotBLock systems
  • Maximum heat transfer with flat-bottom, borosilicate glassware
  • Ideal for reflux and digestion methods
Teardrop stoppers are ideal for the capture and condesation of digestion fumes, preventing cross contamination (sold separately).
$25.10 - $69.40USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Description
Availability Pricing
COMPARE
50 Threaded Digestion Tube, EA
$69.40USD / Each
COMPARE
50 Non-Threaded Digestion Tube, EA
$25.10USD / Each