Environmental Express® SimpleWater BOD Nutrient Buffer Solution
No mess--simply pour contents into your container and add water
  • Range of five standard concentrations to make a volume of 300 mL to 19 L
  • Borosilicate glass vials with leakproof screw caps
BOD dilution water is composed of a variety of nutrients that provide optimal growth media for bacteria in BOD tests. SimpleWater nutrient buffer solutions are concentrates of the four nutrient solutions conveniently combined into one vial. Simply dilute the contents of vial to the specified volume, and you've got your BOD dilution water!
Available in five standard concentrations, borosilicate glass vials with leakproof screwcaps. Vials in pack of 36.
$23.80 - $68.80USD / Pkg of 36
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
300 BOD: SimpleWater (Nutrient Buffer Solution), 300mL, 36/Box
$23.80USD / Pkg of 36
COMPARE
3000 BOD: SimpleWater (Nutrient Buffer Solution), 3L, 36/Box
$51.20USD / Pkg of 36
COMPARE
4000 BOD: SimpleWater (Nutrient Buffer Solution), 4L, 36/Box
$52.30USD / Pkg of 36
COMPARE
6000 BOD: SimpleWater (Nutrient Buffer Solution), 6L, 36/Box
$36.40USD / Pkg of 36
COMPARE
19000 BOD: SimpleWater (Nutrient Buffer Solution), 19L, 36/Box
$68.80USD / Pkg of 36