Environmental Express® SimpleDist® Complete Distillation Systems
Safe and easy distillation system – the name says it all!
  • Conform to EPA and Standard Methods
  • System reduces labor and clean-up time
  • Unique design replaces much of the fragile glassware usually involved in such procedures
The optimal system for performing cyanide, phenol, and ammonia distillations following established EPA methods 335.4 and 350.1, and Standard Methods 4500-CN-C and 4500-NH3.
The distillate is collected in a disposable trap, and does not require the use of a chiller or water lines. The sample boils in the glass boiling tube, and the gas is pulled by vacuum into the trap where it is collected. A hydrophobic porous frit in the trap prevents solution from flowing back into the boiling tube. When complete, the trap is removed and the contents are ready for analysis — Simple!
System comes complete with everything for analysis.
$8,129.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Operating Temperature (° C) Power (VAC) Tube Size (mL)
Availability Pricing
COMPARE
150 120 50
$8,129.00USD / Each
COMPARE
150 240 50
$8,129.00USD / Each