Environmental Express® Radiological Extraction Disposable Filter Funnels
Prevent sample-to-sample contamination to ensure accurate alpha particle spectra
  • Polypropylene construction withstands exposure to acids and bases
  • Choose convenient preassembled filter units or separate funnels and filters to accommodate any protocol
Total funnel capacity is 68 mL and filter diameter is 25 mm. Funnel stem fits a standard rubber stopper hole. Easily remove the filter to analyze filtrate.
$120.50 - $880.00USD / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Sample Volume (mL) Description
Availability Pricing
COMPARE
25 68 Assembled Filter Funnel, PP, 0.1 um, 50/pk
$120.50USD / Pkg of 1
COMPARE
25 68 Assembled Filter Funnel, PP, 0.1 um, 250/pk
$560.00USD / Pkg of 1
COMPARE
25 68 Assembled Filter Funnel, PP and PVDF, 0.45 um, 250/pk
$880.00USD / Pkg of 1
COMPARE
68 Unassembled Filter Funnel, membrane not included, 400/pk
$344.25USD / Pkg of 1