Environmental Express® HotBlock Accessories

Temperature probes and timers enhance your hot blocks
  • Temperature probes fit 15, 50, and 100 mL digestion cups
Make digestion easier with these accessories for the Environmental Express HotBlocks.
$187.50 - $231.75USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Temperature Probe, with PTFE Holder, Fits 50 mL Digestion Tube
$231.75USD / Each
COMPARE
Temperature Probe, with PTFE Holder, Fits 100 mL Digestion Tube
$226.75USD / Each
COMPARE
Temperature Probe, 150°C, Fits 15 mL Digestion Tube
$226.75USD / Each
COMPARE
Programmable Digestion Timer, 120 V
$187.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat