EMD Millipore Air Sampler Agar Strips
Choose from strips with or without media
Compatible with the Biotest Hycon RCS® Air Sampler 86509-00, the sampler operates on impaction principle, drawing airborne microorganisms onto an agar media strip for later incubation and analysis.
$168.75 - $195.00USD / Box of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
EMD Millipore HYCON® 941605 Blank Agar Strips; 50/Box EW-86509-60 Mfr # 1441070050
COMPARE
Biotest Hycon RCS Air Sampler Blank Agar Strips; 50/Box
$168.75USD / Box of 50
EMD Millipore HYCON® 941225 DG18 Agar Strips; 25/Box EW-86509-62 Mfr # 1442450025
COMPARE
Biotest Hycon RCS Air Sampler DG18 Agar Strips; 25/Box
$195.00USD / Box of 25