Drum and Barrel Heavy-Duty Electric Motor Mixers
Heavy-duty mixers for mixing contents of 55-gallon drums and barrels—choose from three mounting styles
These high-quality mixers are available in three mounting versions for heavy-duty mixing. Adjustable hand screws secure the drum-lip model to the rim of 55-gallon (200 liter) drums. The C-clamp model mounts to any open head tank or drum and allows you to adjust the angle of the mixer. The 2" NPT(M) mount model screws directly into a standard 2" (51 mm) female bung to keep odors in and debris out.
Drum-lip and NPT(M) mount models include a folding impeller to pass through bung that attaches to the shaft with set screw and opens to a maximum diameter of 33⁄4" (95 mm) during operation. C-clamp models include a 4" (102 mm) fixed impeller, and are intended for use with an open tank. All mixers deliver maximum speed of 1725 rpm.
$1,442.00 - $1,765.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Torque (in-oz) Motor hp Mounting Type
Availability Pricing
Neptune Drum/barrel heavy-duty mixer with C-clamp; 1/4 hp, 115/230 VAC EW-04318-15 Mfr # B1.0
COMPARE
144 1/4 C-clamp
$1,442.00USD / Each
Neptune Drum/barrel heavy-duty mixer with drum lip mounting; 1/2 hp, 115/230 VAC EW-04318-20 Mfr # F-3.0
COMPARE
288 1/2 Drum-lip clamp
$1,765.00USD / Each
Neptune Drum/barrel heavy-duty mixer with 2" NPT(M) mounting; 1/4 hp, 115/230 VAC EW-50500-00 Mfr # E-1.0
COMPARE
144 1/4 2\" NPT(M)
$1,575.00USD / Each
Neptune Drum/barrel heavy-duty mixer with 2" NPT(M) mounting; 1/2 hp, 115/230 VAC EW-50500-10 Mfr # E-3.0
COMPARE
288 1/2 2\" NPT(M)
$1,723.00USD / Each